Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-33 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$207.00 - $390.00
Specifications
Antigen | IL-33 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 0.5-2 ug/ml, Simple Western 1:200 |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224858SS
![]() |
Novus Biologicals
NBP224858SS |
0.025 mg |
Each for $207.00
|
|
|||||
NBP224858
![]() |
Novus Biologicals
NBP224858 |
0.1 mg |
Each for $390.00
|
|
|||||
Description
IL-33 Polyclonal specifically detects IL-33 in Human, Canine, Monkey samples. It is validated for Western Blot, Simple Western.Specifications
IL-33 | |
Western Blot 0.5-2 ug/ml, Simple Western 1:200 | |
Polyclonal | |
Rabbit | |
Human, Canine, Monkey | |
O95760 | |
90865 | |
A portion of amino acids 50-100 of human IL-33 was used as the immunogen for this antibody. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
0.5 mg/mL | |
Western Blot | |
Unconjugated | |
RUO | |
0.2 ml PBS and 0.2% Gelatin with 0.05% Sodium Azide | |
C9orf26chromosome 9 open reading frame 26 (NF-HEV), DKFZp586H0523, DVS27, DVS27-related protein, IL1F11, IL-1F11, IL-33, interleukin 33, Interleukin-1 family member 11, interleukin-33, NFHEV, NF-HEVNFEHEV, Nuclear factor from high endothelial venules, RP11-575C20.2 | |
IL33 | |
IgG | |
Affinity Purified | |
100% homologous in human (isoforms CRA_a and CRA_b). |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title