Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-34 Antibody (36W18), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP221982A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP221982A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP221982A32 Supplier Novus Biologicals Supplier No. NBP221982AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-34 Monoclonal specifically detects IL-34 in Human samples. It is validated for ELISA.

Specifications

Antigen IL-34
Applications ELISA
Classification Monoclonal
Clone 36W18
Conjugate Alexa Fluor 532
Dilution ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias C16orf77, IL-34, interleukin 34
Gene Symbols IL34
Host Species Mouse
Immunogen Human recombinant IL-34
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 146433
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.