Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-34 Antibody (578416), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB347387 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB347387 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB347387 Supplier Novus Biologicals Supplier No. FAB5265MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-34 Monoclonal antibody specifically detects IL-34 in Human samples. It is validated for Neutralization, Flow Cytometry, CyTOF

Specifications

Antigen IL-34
Applications Neutralization, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 578416
Conjugate mFluor Violet 450 SE
Dilution Neutralization, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias C16orf77, IL-34, interleukin 34
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human IL-34, Asn21-Pro242, Accession # Q6ZMJ4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 146433
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.