Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-36 beta/IL-1F8 Antibody (MM0387-2Q27), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP211687A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP211687A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP211687A70 Supplier Novus Biologicals Supplier No. NBP211687AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-36 beta/IL-1F8 Monoclonal specifically detects IL-36 beta/IL-1F8 in Human samples. It is validated for Western Blot.

Specifications

Antigen IL-36 beta/IL-1F8
Applications Western Blot
Classification Monoclonal
Clone MM0387-2Q27
Conjugate Alexa Fluor 750
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias family of interleukin 1-eta, FIL1, FIL1 eta, FIL1-(ETA), FIL1H, FILI-(ETA), IL-1 eta, IL1-ETA, IL1F8, IL-1F8, IL1F8 (Canonical product IL-1F8a), IL-1F8 (FIL1-eta), IL1H2, IL-1H2, interleukin 1 family, member 8 (eta), interleukin 1, eta, Interleukin 36, Beta, Interleukin-1 eta, interleukin-1 family member 8, Interleukin-1 homolog 2, Interleukin-1 Superfamily e, Interleukin-36 Beta
Gene Symbols IL36B
Host Species Mouse
Immunogen Human recombinant IL-36 beta/IL-1F8
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 27177.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.