Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-36 beta/IL-1F8 Antibody (MM0387-2Q27), HRP, Novus Biologicals™
SDP

Catalog No. NBP211687H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP211687H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP211687H Supplier Novus Biologicals Supplier No. NBP211687H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-36 beta/IL-1F8 Monoclonal specifically detects IL-36 beta/IL-1F8 in Human samples. It is validated for Western Blot.

Specifications

Antigen IL1F8
Applications Western Blot
Classification Monoclonal
Clone MM0387-2Q27
Conjugate HRP
Dilution Western Blot
Formulation PBS with No Preservative
Gene Alias family of interleukin 1-eta, FIL1, FIL1 eta, FIL1-(ETA), FIL1H, IL-1 eta, IL1F8 (Canonical product IL-1F8a), IL-1F8 (FIL1-eta), IL-1F8IL1-ETA, IL-1H2MGC126880, IL1H2MGC126882, interleukin 1 family, member 8 (eta), interleukin 1, eta, Interleukin-1 eta, interleukin-1 family member 8, Interleukin-1 homolog 2, Interleukin-1 Superfamily e
Gene Symbols IL36B
Host Species Mouse
Immunogen Human recombinant IL-36 beta/IL-1F8
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 27177
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.