Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-36 gamma/IL-1F9 Antibody (Jussy-1), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB120644 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB120644 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB120644 Supplier Novus Biologicals Supplier No. NBP311676AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-36 gamma/IL-1F9 Monoclonal specifically detects IL-36 gamma/IL-1F9 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen IL-36 gamma/IL-1F9
Applications Western Blot, ELISA
Classification Monoclonal
Clone Jussy-1
Conjugate Alexa Fluor 594
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias IL-1 epsilon, IL-1 Related Protein 2, IL-1(EPSILON), IL1E, IL-1-epsilon, IL1F9, IL-1F9, IL1H1, IL-1H1, IL-1-Related Protein 2, IL1RP2, IL-1RP2, interleukin 1 family, member 9, interleukin 1-related protein 2, Interleukin 36, Gamma, Interleukin-1 epsilon, interleukin-1 family member 9, Interleukin-1 homolog 1, Interleukin-36 Gamma
Host Species Mouse
Immunogen Recombinant human IL-36 gamma/IL-1F9.
Purification Method >95%, by SDS-PAGE
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 56300
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.