Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-36 gamma/IL-1F9 Antibody (OTI2F4), Novus Biologicals™
SDP

Catalog No. NBP245749 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP245749 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP245749 Supplier Novus Biologicals Supplier No. NBP245749
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in Western Blot, Immunohistochemistry, Immunofluorescence

IL-36 gamma/IL-1F9 Monoclonal specifically detects IL-36 gamma/IL-1F9 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence.

Specifications

Antigen IL1F9
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI2F4
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:4000, Immunohistochemistry 1:150, Immunofluorescence 1:100
Formulation PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide
Gene Alias IL1E, IL-1F9IL-1 epsilon, IL1H1IL-1-epsilon, IL-1H1IL-1-related protein 2, IL1RP2IL-1 related protein 2, IL-1RP2IL-1(EPSILON), interleukin 1 family, member 9, interleukin 1-related protein 2, Interleukin-1 epsilon, interleukin-1 family member 9, Interleukin-1 homolog 1
Gene Symbols IL36G
Host Species Mouse
Immunogen Full length human recombinant protein of human IL1F9(NP_062564) produced in HEK293T cell.
Molecular Weight of Antigen 18.5 kDa
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56300
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.