Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-36 gamma/IL-1F9 Rabbit anti-Human, Clone: 13, Novus Biologicals™
SDP

Catalog No. NB020046 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB020046 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB020046 Supplier Novus Biologicals Supplier No. NBP289281100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IL-36 gamma/IL-1F9 Monoclonal specifically detects IL-36 gamma/IL-1F9 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen IL-36 gamma/IL-1F9
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 13
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:10000-1:20000
Formulation 0.2 um filtered solution in PBS
Gene Alias IL-1 epsilon, IL-1 Related Protein 2, IL-1(EPSILON), IL1E, IL-1-epsilon, IL1F9, IL-1F9, IL1H1, IL-1H1, IL-1-Related Protein 2, IL1RP2, IL-1RP2, interleukin 1 family, member 9, interleukin 1-related protein 2, Interleukin 36, Gamma, Interleukin-1 epsilon, interleukin-1 family member 9, Interleukin-1 homolog 1, Interleukin-36 Gamma
Host Species Rabbit
Immunogen Recombinant Human IL-36 gamma/IL-1F9 Protein
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 56300
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.