Learn More
Description
Specifications
Specifications
| Antigen | IL-36Ra/IL-1F5 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 700 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | FIL1, FIL1 delta, FIL1-delta, IL-1RP3, IL-1L1, IL-1-delta), FIL1DIL-1 delta, IL1F5 (Canonical product IL-1F5a), IL-1RP3, IL36RA, IL-36RA, interleukin 1 family, member 5 (delta), interleukin 1, delta, Interleukin 36 Receptor Antagonist, interleukin-1 family member 5, Interleukin-1 HY1, Interleukin-1 receptor antagonist homolog 1, Interleukin-1-like protein 1, Interleukin-36 Receptor Antagonist |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against a 19 amino acid peptide near the carboxy terminus of human IL-36RN. The immunogen is located within the last 50 amino acids of IL-36RN. Amino Acid Squence: PEADQPVRLTQLPENGGWN |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
