Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-37 Monoclonal Antibody (37D12), PE, eBioscience™
GREENER_CHOICE

Catalog No. 501122391
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-2391 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-112-2391 Supplier Invitrogen™ Supplier No. 12737942
Only null left
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

Description: This 37D12 monoclonal antibody reacts with human IL-37. IL-37 is a member of the interleukin 1 cytokine family and is also known as IL-1F7 and IL-1H4. IL-37 suppresses innate inflammatory and immune responses and exhibits anti-inflammatory characteristics resembling those of IL-1R8. IL-37 binds to the alpha chain of IL-18R with low affinity and does not recruit the essential signaling co-receptor IL-18 beta or mediate pro-inflammatory activity. IL-37 also binds to IL-18 binding protein (IL-18BP) and enhances the antagonistic effects of IL-18BP. IL-37 forms a trimeric complex with IL-18BP and IL-18R beta, which results in the blockade of IL-18 activity. Transcripts of IL-37 have been detected in human tissues, such as lung, testis, and colon tumors, as well as some human cell lines. Applications Reported: This 37D12 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This 37D12 antibody has been pre-titrated and tested by intracellular staining followed by flow cytometric analysis of normal human peripheral blood cells using the Intracellular Fixation & Permeabilization Buffer Set (Product # 88-8824-00) and protocol. Please refer to BestProtocols®: Protocol A: Two step protocol for (cytoplasmic) intracellular proteins located under the Resources Tab online. This can be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sampl...

The protein encoded by this gene is a member of the interleukin 1 cytokine family. This cytokine can bind to, and may be a ligand for interleukin 18 receptor (IL18R1/IL-1Rrp). This cytokine also binds to interleukin 18 binding protein (IL18BP), an inhibitory binding protein of interleukin 18 (IL18), and subsequently forms a complex with IL18 receptor beta subunit, and through which it inhibits the activity of IL18. This gene along with eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. Five alternatively spliced transcript variants encoding distinct isoforms have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-37
Applications Flow Cytometry
Classification Monoclonal
Clone 37D12
Concentration 5 μL/Test
Conjugate PE
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene IL37
Gene Accession No. Q9NZH6
Gene Alias FIL1; FIL1 zeta; FIL1(ZETA); FIL1Z; IL-1 zeta; IL1F7; IL-1F7; IL1F7 (canonical product IL-1F7b); IL-1F7b (IL-1H4, IL-1H, IL-1RP1); IL-1H; IL1H4; IL-1H4; IL1RP1; IL-1RP1; IL-1X; IL-1X protein; IL37; IL-37; ILN; Interleukin; interleukin 1 family member 7; interleukin 1, zeta; interleukin 37; Interleukin-1 family member 7; Interleukin-1 homolog 4; interleukin-1 superfamily z; Interleukin-1 zeta; Interleukin-1-related protein; interleukin-23; Interleukin37; Interleukin-37
Gene Symbols IL37
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27178
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.