Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-38/IL-1F10 Antibody (798036R), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB374061 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB374061 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB374061 Supplier Novus Biologicals Supplier No. FAB7774RK
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IL-38/IL-1F10 Monoclonal specifically detects IL-38/IL-1F10 in Mouse samples. It is validated for ELISA, Flow Cytometry, Western Blot.

Specifications

Antigen IL-38/IL-1F10
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 798036R
Conjugate DyLight 488
Dilution Western Blot, Flow Cytometry, ELISA
Formulation 50mM Sodium Borate
Gene Alias interleukin 1 family, member 10 (theta), UNQ6119/PRO20041
Host Species Rat
Immunogen E.coli-derived recombinant mouse IL-38/IL-1F10, Met1-Arg152, Accession # Q8R459
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 84639
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.