Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-3R alpha/CD123 Antibody (001), FITC, Novus Biologicals™
SDP

Catalog No. NB383149 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB383149 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB383149 Supplier Novus Biologicals Supplier No. NBP312772F
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IL-3R alpha/CD123 Monoclonal antibody specifically detects IL-3R alpha/CD123 in Mouse samples. It is validated for ELISA

Specifications

Antigen IL-3R alpha/CD123
Applications ELISA
Classification Monoclonal
Clone 001
Conjugate FITC
Formulation PBS
Gene Alias CD123, CD123 antigen, hIL3Ra, hIL-3Ra, IL-3 receptor subunit alpha, IL3R, IL-3R subunit alpha, IL-3RA, IL-3R-alpha, IL3RAY, IL3RX, IL3RY, interleukin 3 receptor, alpha (low affinity), interleukin-3 receptor subunit alpha, MGC34174
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL-3R alpha/CD123 (Met1-Lys331; Accession#: EDL20630.1)
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline B Cell Development and Differentiation Markers, Cytokine Research, Oncogenes, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 3563
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.