Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-3R alpha/CD123 Antibody (017), HRP
SDP

Catalog No. NB108252 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB108252 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB108252 Supplier Novus Biologicals Supplier No. NBP289550H

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Monoclonal Antibody

IL-3R alpha/CD123 Monoclonal specifically detects IL-3R alpha/CD123 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen IL-3R alpha/CD123
Applications Western Blot, ELISA
Classification Monoclonal
Clone 17
Conjugate HRP
Formulation PBS
Gene Alias CD123, CD123 antigen, hIL3Ra, hIL-3Ra, IL-3 receptor subunit alpha, IL3R, IL-3R subunit alpha, IL-3RA, IL-3R-alpha, IL3RAY, IL3RX, IL3RY, interleukin 3 receptor, alpha (low affinity), interleukin-3 receptor subunit alpha, MGC34174
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-3R alpha/CD123 (NP_002174.1; Met 1-Arg 305).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline B Cell Development and Differentiation Markers, Cytokine Research, Oncogenes, Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 3563
Test Specificity No cross-reactivity in ELISA with: Human IL6R
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.