Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-3R alpha/CD123 Antibody (18N6B12), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP227225G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227225G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP227225G Supplier Novus Biologicals Supplier No. NBP227225G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

IL-3R alpha/CD123 Monoclonal specifically detects IL-3R alpha/CD123 in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen IL-3R alpha/CD123
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone 18N6B12
Conjugate DyLight 488
Dilution Western Blot, Flow Cytometry
Gene Accession No. P26951
Gene Alias CD123, CD123 antigen, hIL3Ra, hIL-3Ra, IL-3 receptor subunit alpha, IL3R, IL-3R subunit alpha, IL-3RA, IL-3R-alpha, IL3RAY, IL3RX, IL3RY, interleukin 3 receptor, alpha (low affinity), interleukin-3 receptor subunit alpha, MGC34174
Gene Symbols IL3RA
Host Species Mouse
Immunogen RERVYEFLSAWSTPQRFEC (aa275-293).
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 3563
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.