Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-3R alpha/CD123 Antibody, Novus Biologicals™
SDP

Catalog No. NBP227160 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227160 0.1 mg
NBP227160SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227160 Supplier Novus Biologicals Supplier No. NBP227160
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IL-3R alpha/CD123 Polyclonal specifically detects IL-3R alpha/CD123 in Human samples. It is validated for Western Blot.

Specifications

Antigen IL-3R alpha/CD123
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 3-5ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. P26951
Gene Alias CD123, CD123 antigen, hIL3Ra, hIL-3Ra, IL-3 receptor subunit alpha, IL3R, IL-3R subunit alpha, IL-3RA, IL-3R-alpha, IL3RAY, IL3RX, IL3RY, interleukin 3 receptor, alpha (low affinity), interleukin-3 receptor subunit alpha, MGC34174
Gene Symbols IL3RA
Host Species Rabbit
Immunogen A portion of amino acids120-170 of human IL-3 R alpha/CD123 was used as the immunogen for this antibody.
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cytokine Research, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 3563
Test Specificity The IL-3Ra amino acid sequence used as immunogen is 100% homologous in human and chimpanzee.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.