Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-3R beta Antibody (130705), HRP, Novus Biologicals™
SDP

Catalog No. NB296097 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB296097 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB296097 Supplier Novus Biologicals Supplier No. FAB5492H
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IL-3R beta Monoclonal antibody specifically detects IL-3R beta in Mouse samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen IL-3R beta
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 130705
Conjugate HRP
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias AIC2A, BetaIl3, Bil3, Csfgmrb, IL3R, IL3RB, Il3rb2
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse IL-3 R beta , His23-Trp440, Accession # P26954
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1439
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.