Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-4 Antibody (101), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB068731 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB068731 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB068731 Supplier Novus Biologicals Supplier No. NBP290001C
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IL-4 Monoclonal specifically detects IL-4 in Human samples. It is validated for ELISA.

Specifications

Antigen IL-4
Applications ELISA
Classification Monoclonal
Clone 101
Conjugate DyLight 650
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias B cell growth factor 1, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL-4B_cell stimulatory factor 1, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-4 (P05112-1; His25-Ser153).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Asthma, Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3565
Test Specificity No cross-reactivity in ELISA with: Mouse IL4 Canine IL4
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.