Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-4 Antibody (11B11), Alexa Fluor™ 350 - Chimeric, Novus Biologicals™
SDP

Catalog No. NP2266A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP2266A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP2266A30 Supplier Novus Biologicals Supplier No. NBP252656AF350
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IL-4 Monoclonal specifically detects IL-4 in Mouse samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry.

Specifications

Antigen IL-4
Applications Flow Cytometry, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 11B11
Conjugate Alexa Fluor 350
Dilution Flow Cytometry, ELISA, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias B cell growth factor 1, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL-4B_cell stimulatory factor 1, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra
Gene Symbols IL4
Host Species Rabbit
Immunogen BSF-1/ IL-4 purified from serum-free supernatants of cultured EL-4 cells (a C57BL/6 thymoma) induced with 4 Beta-phorbol 12 Beta-myristate 12 alpha -acetate for 48h.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3565
Test Specificity Binds to a core functional region of murine IL-4, which prevents this cytokine from interacting with its cognate receptor (Yao et al., 2006 PMID: 16361318).
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.