Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-4 Antibody (99605), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB346942 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB346942 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB346942 Supplier Novus Biologicals Supplier No. FAB6543MFV500

Mouse Monoclonal Antibody

IL-4 Monoclonal antibody specifically detects IL-4 in Porcine samples. It is validated for Western Blot, Flow Cytometry, CyTOF

Specifications

Antigen IL-4
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 99605
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias B cell growth factor 1, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL-4B_cell stimulatory factor 1, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra
Host Species Mouse
Immunogen E. coli-derived recombinant porcine IL-4, His25-Cys133, Accession # Q04745
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Asthma, Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3565
Target Species Pig
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.