Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-4 Antibody (BVD4-1D11), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB10064798Y Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064798Y 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10064798Y Supplier Novus Biologicals Supplier No. NB10064798AF647
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IL-4 Monoclonal specifically detects IL-4 in Mouse samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot.

Specifications

Antigen IL-4
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone BVD4-1D11
Conjugate Alexa Fluor 647
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias B cell growth factor 1, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL-4B_cell stimulatory factor 1, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra
Gene Symbols IL4
Host Species Rat
Immunogen Recombinant Mouse IL-4
Purification Method Protein A or G purified
Quantity 0.25 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Asthma, Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3565
Test Specificity This is specific for mouse Interleukin-4 (IL-4). Interleukin-4 is a potent lymphoid growth factor, which stimulates the growth and survival of certain B and T cells. Clone BVD4-1D11 is reported to neutralise the effects of mouse IL-4 (3).
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.