Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-4 Antibody (V17P1B4*B12), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP261126A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP261126A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP261126A70 Supplier Novus Biologicals Supplier No. NBP261126AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-4 Monoclonal specifically detects IL-4 in Fish samples. It is validated for Western Blot, ELISA.

Specifications

Antigen IL-4
Applications Western Blot, ELISA
Classification Monoclonal
Clone V17P1B4*B12
Conjugate Alexa Fluor 750
Dilution Western Blot, ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias B cell growth factor 1, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL-4B_cell stimulatory factor 1, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra
Gene Symbols IL4
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide- AQAIYSRPHP
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3565
Target Species Fish
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.