Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-4I1 Antibody (1006231), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB303237 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB303237 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB303237 Supplier Novus Biologicals Supplier No. FAB56843N

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rat Monoclonal Antibody

IL-4I1 Monoclonal antibody specifically detects IL-4I1 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF

Specifications

Antigen IL-4I1
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 1006231
Conjugate Alexa Fluor 700
Dilution Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias FIG1, Fig-1 protein, hFIG1, IL4-induced protein 1, interleukin 4 induced 1, interleukin four induced 1, LAAO, L-amino-acid oxidase, LAO, UNQ636/PRO1265
Host Species Rat
Immunogen Chinese Hamster Ovary cell line, CHO-derived human IL-4I1, Met1-His567, Accession # Q96RQ9
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 259307
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.