Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-4R alpha Antibody (25463), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB365632 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB365632 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB365632 Supplier Novus Biologicals Supplier No. FAB230L
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-4R alpha Monoclonal antibody specifically detects IL-4R alpha in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Neutralization, Immunocytochemistry, CyTOF

Specifications

Antigen IL-4R alpha
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Neutralization, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 25463
Conjugate DyLight 550
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Neutralization, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD124 antigen, IL-4 receptor subunit alpha, IL-4R subunit alpha, IL-4RA, IL4RACD124, IL-4R-alpha, interleukin 4 receptor, interleukin-4 receptor alpha chain, interleukin-4 receptor subunit alpha
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-4 R, Gly24-His232, Accession # P24394
Purification Method Protein A or G purified from ascites
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Asthma, B Cell Development and Differentiation Markers, Cancer, Immunology, Myeloid derived Suppressor Cell
Primary or Secondary Primary
Gene ID (Entrez) 3566
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.