Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-5 Antibody (TRFK5), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB296338 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB296338 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB296338 Supplier Novus Biologicals Supplier No. FAB405G

Rat Monoclonal Antibody

IL-5 Monoclonal antibody specifically detects IL-5 in Human, Mouse samples. It is validated for ELISA Standard (Matched Pair), Neutralization, ELISA (Matched Antibody Pair), ELISA (Matched Antibody Pair), Flow Cytometry, CyTOF-reported

Specifications

Antigen IL-5
Applications ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Flow Cytometry
Classification Monoclonal
Clone TRFK5
Conjugate Alexa Fluor 488
Dilution ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Intracellular Staining by Flow Cytometry, CyTOF-reported
Formulation 50mM Sodium Borate
Gene Alias B-cell differentiation factor I, EDF, Eosinophil differentiation factor, IL-5T-cell replacing factor, interleukin 5 (colony-stimulating factor, eosinophil), interleukin-5, TRFB cell differentiation factor I
Host Species Rat
Immunogen Mouse T cell clone-derived partially purified recombinant mouse IL-5
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Asthma, Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3567
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.