Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-5 Mouse anti-Bovine, Goat, Sheep, Clone: MT52B34, Novus Biologicals™
SDP

Catalog No. NB171902 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1000 μg
250 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB171902 250 μg
NB171901 1000 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB171902 Supplier Novus Biologicals Supplier No. NBP318278250UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-5 Monoclonal antibody specifically detects IL-5 in Bovine, Goat, Sheep samples. It is validated for ELISA, ELISPOT

Specifications

Antigen IL-5
Applications ELISA
Classification Monoclonal
Clone MT52B34
Concentration 0.5 mg/ml
Conjugate Unconjugated
Formulation 0.2 um filtered PBS
Gene Alias B-cell differentiation factor I, EDF, Eosinophil differentiation factor, IL-5T-cell replacing factor, interleukin 5 (colony-stimulating factor, eosinophil), interleukin-5, TRFB cell differentiation factor I
Host Species Mouse
Immunogen Recombinant bovine IL-5
Purification Method Protein G purified
Quantity 250 μg
Regulatory Status RUO
Research Discipline Asthma, Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3567
Target Species Bovine, Goat, Sheep
Content And Storage Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.