Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-5R alpha/CD125 Antibody (26815), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB175555 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB175555 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB175555 Supplier Novus Biologicals Supplier No. FAB253D
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-5R alpha/CD125 Monoclonal antibody specifically detects IL-5R alpha/CD125 in Human samples. It is validated for Western Blot, Flow Cytometry

Specifications

Antigen IL-5R alpha/CD125
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone 26815
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias CD125 antigen, CDw125CD125, HSIL5R3, IL-5 receptor subunit alpha, IL5R, IL-5R subunit alpha, IL-5RA, IL-5R-alpha, interleukin 5 receptor type 3, interleukin 5 receptor, alpha, interleukin-5 receptor alpha chain, interleukin-5 receptor alpha subunit, interleukin-5 receptor subunit alpha, MGC26560
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-5 R alpha/CD125, Asp21-Arg335
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3568
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.