Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-6 Antibody (1936R), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB349315 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB349315 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB349315 Supplier Novus Biologicals Supplier No. FAB2061RMFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-6 Monoclonal specifically detects IL-6 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), Neutralization.

Specifications

Antigen IL-6
Applications Flow Cytometry (Intracellular Staining), Neutralization
Classification Monoclonal
Clone 1936R
Conjugate mFluor Violet 450 SE
Dilution Neutralization, Intracellular Staining by Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6
Host Species Mouse
Immunogen E. coli-derived recombinant human IL-6, Val30-Met212, Accession # P05231
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Biologically Active Proteins, Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 3569
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.