Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-6 Antibody (B-E8), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB388457 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB388457 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB388457 Supplier Novus Biologicals Supplier No. NBP314588APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-6 Monoclonal antibody specifically detects IL-6 in Human samples. It is validated for Flow Cytometry, ELISA, Functional Assay, Enzyme-linked Immunosorbent Spot

Specifications

Antigen IL-6
Applications Flow Cytometry, ELISA, Functional Assay, ELISpot
Classification Monoclonal
Clone B-E8
Conjugate APC
Formulation PBS
Gene Alias B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6
Host Species Mouse
Immunogen Recombinant human IL-6
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Biologically Active Proteins, Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 3569
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.