Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EBI3 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP203944 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP203944 0.1 mg
NBP203944SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP203944 Supplier Novus Biologicals Supplier No. NBP203944
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EBI3 Polyclonal specifically detects EBI3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen IL-27/IL-35 EBI3 Subunit
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 0.5-2 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. NP_056581.1
Gene Alias cytokine receptor, EBV-induced gene 3 protein, Epstein-Barr virus induced 3, Epstein-Barr virus induced gene 3, Epstein-Barr virus-induced gene 3 protein, IL-27 subunit beta, IL27B, IL-27B, interleukin-27 subunit beta
Gene Symbols EBI3
Host Species Rabbit
Immunogen A portion of amino acids 180-228 of mouse EBI3 was used as the immunogen.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Chemokine Research, Cytokine Research, Hypoxia, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10148
Test Specificity Reacts with Human, Mouse and Rat. The amino acid sequence used as immunogen is 100% homologous in mouse (isoforms CRA_a and CRA_b) and rat and 72% homologous in human, dog and rhesus monkey.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.