Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EBI3 Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.50 - $438.00

Specifications

Antigen IL-27/IL-35 EBI3 Subunit
Concentration 0.5 mg/ml
Dilution Western Blot 0.5-2 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP203944SS
SDP
View Documents
Novus Biologicals
NBP203944SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP203944
SDP
View Documents
Novus Biologicals
NBP203944
0.1 mg
Each for $438.00
Only null left
Add to Cart
 
Description

Description

EBI3 Polyclonal specifically detects EBI3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

IL-27/IL-35 EBI3 Subunit
Western Blot 0.5-2 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml
Polyclonal
Rabbit
Apoptosis, Cancer, Chemokine Research, Cytokine Research, Hypoxia, Immunology, Innate Immunity, Signal Transduction
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
cytokine receptor, EBV-induced gene 3 protein, Epstein-Barr virus induced 3, Epstein-Barr virus induced gene 3, Epstein-Barr virus-induced gene 3 protein, IL-27 subunit beta, IL27B, IL-27B, interleukin-27 subunit beta
EBI3
IgG
Affinity Purified
Reacts with Human, Mouse and Rat. The amino acid sequence used as immunogen is 100% homologous in mouse (isoforms CRA_a and CRA_b) and rat and 72% homologous in human, dog and rhesus monkey.
0.5 mg/ml
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human, Mouse, Rat
NP_056581.1
10148
A portion of amino acids 180-228 of mouse EBI3 was used as the immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.