Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-6 Antibody, Novus Biologicals™
SDP

Catalog No. NB6001131 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.04 mL
0.2 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB6001131 0.2 mL
NB6001131SS 0.04 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB6001131 Supplier Novus Biologicals Supplier No. NB6001131
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 39 publications

IL-6 Polyclonal specifically detects IL-6 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockdown Validated.

Specifications

Antigen IL-6
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), KnockDown, KnockDown
Classification Polyclonal
Concentration 77 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA 1:1000-1:5000, Immunohistochemistry 1:400-1:800, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:400-1:800, Immunohistochemistry-Paraffin 1:400 - 1:800, Immunohistochemistry-Frozen, Knockdown Validated
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with No Preservative
Gene Accession No. P05231
Gene Alias B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6
Gene Symbols IL6
Host Species Rabbit
Immunogen This whole rabbit serum was prepared by repeated immunizations with recombinant human IL-6 produced in E.coli. (Uniprot: P05231)
Molecular Weight of Antigen 23.72 kDa
Purification Method Unpurified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3569
Test Specificity Anti-IL-6 antiserum detects recombinant and native IL-6 present in body fluids and cell supernatants in various assays (ie. IL-1 stimulated IL-6 production from fibroblasts).
Target Species Human, Mouse, Rat, Monkey
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.