Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-6 Rabbit anti-Rat, Clone: 7, Novus Biologicals™
SDP

Catalog No. NB025554 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB025554 100 μg
NB025555 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB025554 Supplier Novus Biologicals Supplier No. NBP290873100UL

Rabbit Monoclonal Antibody

IL-6 Monoclonal specifically detects IL-6 in Rat samples. It is validated for ELISA.

Specifications

Antigen IL-6
Applications ELISA
Classification Monoclonal
Clone 7
Conjugate Unconjugated
Dilution ELISA 1:5000-1:10000
Formulation 0.2 um filtered solution in PBS
Gene Alias B cell stimulatory factor-2, B-cell differentiation factor, BSF-2, BSF2CTL differentiation factor, CDF, HGFHSFIFNB2Hybridoma growth factor, IFN-beta-2, IL-6B-cell stimulatory factor 2, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6
Host Species Rabbit
Immunogen Recombinant Rat IL-6 Protein
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Biologically Active Proteins, Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 3569
Target Species Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.