Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-6R alpha Antibody (033), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB039768 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB039768 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB039768 Supplier Novus Biologicals Supplier No. NBP290512C
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IL-6R alpha Monoclonal specifically detects IL-6R alpha in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen IL-6R alpha
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone 33
Conjugate DyLight 650
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias CD126, gp80, IL6Q, IL6R, IL-6R-1, IL6RA, IL-6RA, IL6RQ, interleukin 6 receptor
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL-6R alpha (NP_034689.2; Ile44-Glu357).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3570
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.