Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-6R alpha Antibody (Hu137) [DyLight 680], Novus Biologicals™
SDP

Catalog No. NB401795 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB401795 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB401795 Supplier Novus Biologicals Supplier No. FAB10346Y
Add to Cart
Add to Cart

Human Monoclonal Antibody

IL-6R alpha Monoclonal antibody specifically detects IL-6R alpha in Human samples. It is validated for Flow Cytometry, Neutralization, CyTOF

Specifications

Antigen IL-6R alpha
Applications Flow Cytometry, Neutralization, CyTOF
Classification Monoclonal
Clone Hu137
Conjugate DyLight 680
Dilution Flow Cytometry, Neutralization, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD126, gp80, IL6Q, IL6R, IL-6R-1, IL6RA, IL-6RA, IL6RQ, interleukin 6 receptor
Host Species Human
Immunogen Human IL-6R alpha
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3570
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.