Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-7 Antibody (7417.111), Novus Biologicals™
SDP

Catalog No. NB600837 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600837 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600837 Supplier Novus Biologicals Supplier No. NB600837

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

IL-7 Monoclonal specifically detects IL-7 in Human samples. It is validated for Western Blot, ELISA, Block/Neutralize.

Specifications

Antigen IL-7
Applications Western Blot, ELISA, Blocking Assay
Classification Monoclonal
Clone 7417.111
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, ELISA 1-2 ug/ml, Blocking/Neutralizing 50-200 molar excess
Gene Alias IL-7interleukin-7, interleukin 7
Gene Symbols IL7
Host Species Mouse
Immunogen Recombinant full length human IL7 protein.
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Cytokine Research, Hematopoietic Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3574
Test Specificity The has the ability to neutralize the biological activity of recombinant human IL-7. It may also be used as a capture in a human IL-7 ELISA.
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.