Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-7R alpha/CD127 Antibody (1140A) [DyLight 405], Novus Biologicals™
SDP

Catalog No. NB374587 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB374587 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB374587 Supplier Novus Biologicals Supplier No. FAB7473E
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IL-7R alpha/CD127 Monoclonal specifically detects IL-7R alpha/CD127 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen IL-7R alpha/CD127
Applications Flow Cytometry
Classification Monoclonal
Clone 1140A
Conjugate DyLight 405
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias CD127 antigen, CD127ILRA, CDW127, IL-7 receptor subunit alpha, IL-7R subunit alpha, IL7RA, IL-7RA, IL-7R-alpha, interleukin 7 receptor, interleukin 7 receptor alpha chain, interleukin 7 receptor isoform H5-6, interleukin-7 receptor subunit alpha
Host Species Rabbit
Immunogen NSO mouse myeloma cell line transfected with mouse IL-7 R alpha/CD127, Glu21-Asp239, Accession # P16872
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3575
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.