Learn More
Description
Specifications
Specifications
| Antigen | IL-7R alpha/CD127 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 115 |
| Conjugate | HRP |
| Dilution | ELISA |
| Formulation | PBS |
| Gene Alias | CD127 antigen, CD127ILRA, CDW127, IL-7 receptor subunit alpha, IL-7R subunit alpha, IL7RA, IL-7RA, IL-7R-alpha, interleukin 7 receptor, interleukin 7 receptor alpha chain, interleukin 7 receptor isoform H5-6, interleukin-7 receptor subunit alpha |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL-7R alpha/CD127 (NP_032398.3; Met1-Asp239). |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
