Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-7R alpha/CD127 Antibody (BLR177J), Novus Biologicals™
SDP

Catalog No. NB188089 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB188089 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB188089 Supplier Novus Biologicals Supplier No. NBP314749
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

IL-7R alpha/CD127 Monoclonal antibody specifically detects IL-7R alpha/CD127 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation

Specifications

Antigen IL-7R alpha/CD127
Applications Western Blot, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone BLR177J
Conjugate Unconjugated
Formulation Borate Buffered Saline (BBS) pH 8.2 with, BSA Free with 0.09% Sodium Azide
Gene Alias CD127 antigen, CD127ILRA, CDW127, IL-7 receptor subunit alpha, IL-7R subunit alpha, IL7RA, IL-7RA, IL-7R-alpha, interleukin 7 receptor, interleukin 7 receptor alpha chain, interleukin 7 receptor isoform H5-6, interleukin-7 receptor subunit alpha
Host Species Rabbit
Immunogen residues 21-236 (ECD)
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3575
Target Species Human
Content And Storage Store at 2 - 8°C/ 1 year from date of receipt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.