Learn More
IL-7R alpha/CD127, Rabbit anti-Human, Clone: YIL7R323, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | IL-7R alpha/CD127 |
| Applications | Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Clone | YIL7R323 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry, ELISA |
| Formulation | PBS |
| Gene Alias | CD127 antigen, CD127ILRA, CDW127, IL-7 receptor subunit alpha, IL-7R subunit alpha, IL7RA, IL-7RA, IL-7R-alpha, interleukin 7 receptor, interleukin 7 receptor alpha chain, interleukin 7 receptor isoform H5-6, interleukin-7 receptor subunit alpha |
| Gene Symbols | IL7R |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.