Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-9 Antibody (D9302C12), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB153030 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB153030 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB153030 Supplier Novus Biologicals Supplier No. NBP312011UV
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

IL-9 Monoclonal antibody specifically detects IL-9 in Mouse samples. It is validated for Flow Cytometry

Specifications

Antigen IL-9
Applications Flow Cytometry
Classification Monoclonal
Clone D9302C12
Conjugate DyLight 350
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias Cytokine P40, HP40, IL-9homolog of mouse T cell and mast cell growth factor 40, interleukin 9, interleukin-9, P40, p40 cytokine, p40 T-cell and mast cell growth factor, T-cell growth factor P40
Host Species Armenian Hamster
Immunogen This antibody was raised by immunising Armenian hamsters with Baculovirus-expressed, recombinant murine IL-9.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Asthma, Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3578
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.