Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-9 Antibody (MH9A4), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB153289 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB153289 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB153289 Supplier Novus Biologicals Supplier No. NBP312022JF549
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IL-9 Monoclonal antibody specifically detects IL-9 in Human samples. It is validated for Flow Cytometry, ELISA

Specifications

Antigen IL-9
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone MH9A4
Conjugate Janelia Fluor 549
Dilution Flow Cytometry, ELISA
Formulation 50mM Sodium Borate
Gene Alias Cytokine P40, HP40, IL-9homolog of mouse T cell and mast cell growth factor 40, interleukin 9, interleukin-9, P40, p40 cytokine, p40 T-cell and mast cell growth factor, T-cell growth factor P40
Host Species Rabbit
Immunogen This antibody was raised by immunising C57Bl/6 mice with human IL-9 coupled to ovalbumin.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Asthma, Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3578
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.