Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-9R Antibody (33423), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB175588 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB175588 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB175588 Supplier Novus Biologicals Supplier No. FAB290E
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-9R Monoclonal specifically detects IL-9R in Human samples. It is validated for CyTOF, Flow Cytometry, Neutralization.

Specifications

Antigen IL-9R
Applications CyTOF, Flow Cytometry, Neutralization
Classification Monoclonal
Clone 33423
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias CD129, CD129 antigen, IL-9 receptor, IL-9R, interleukin 9 receptor, interleukin-9 receptor
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-9 R
Purification Method Protein A or G purified from ascites
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 3581
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.