Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-9R Antibody (AH9R2), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB153529 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB153529 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB153529 Supplier Novus Biologicals Supplier No. NBP312033G
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IL-9R Monoclonal antibody specifically detects IL-9R in Human samples. It is validated for Flow Cytometry, Immunofluorescence

Specifications

Antigen IL-9R
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone AH9R2
Conjugate DyLight 488
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias CD129, CD129 antigen, IL-9 receptor, IL-9R, interleukin 9 receptor, interleukin-9 receptor
Host Species Rabbit
Immunogen This antibody was raised by immunising DBA/2 mice with P815 mastocytoma cells transfected with human IL-9R cDNA.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3581
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.