Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-9R Antibody (RM0115-12F43) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB018440 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018440 0.025 mg
NBP122493 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018440 Supplier Novus Biologicals Supplier No. NBP1224930.025MG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IL-9R Monoclonal specifically detects IL-9R in Mouse samples. It is validated for Western Blot.

Specifications

Antigen IL-9R
Applications Western Blot
Classification Monoclonal
Clone RM0115-12F43
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Alias CD129, CD129 antigen, IL-9 receptor, IL-9R, interleukin 9 receptor, interleukin-9 receptor
Gene Symbols IL9R
Host Species Rat
Immunogen Mouse recombinant protein of the extracellular domain of IL-9 R
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3581
Test Specificity This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein of the extracellular domain of IL-9 R.
Reconstitution Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml.
Target Species Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.