Learn More
Invitrogen™ IL10RB Monoclonal Antibody (90220)

Description
In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-10 R alpha, rhIFN-gamma RI, and rhIFN-gamma R2 is observed. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
Specifications
Specifications
| Antigen | IL10RB |
| Applications | Flow Cytometry, Neutralization, Western Blot |
| Classification | Monoclonal |
| Clone | 90220 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | IL10RB |
| Gene Accession No. | Q08334 |
| Gene Alias | 6620401D04Rik; AI528744; CDW210B; CRF2-4; CRFB4; cytokine receptor class-II CRF2-4; cytokine receptor class-II member 4; Cytokine receptor family 2 member 4; cytokine receptor family II, member 4; D16H21S58; D21S58; D21S58h; D21S66; IL-10 receptor subunit beta; IL-10R subunit 2; IL-10R subunit beta; Il10r2; IL-10R2; IL10RB; IL-10RB; inte; interleukin 10 receptor 2; interleukin 10 receptor subunit beta; interleukin 10 receptor, beta; interleukin-10 receptor subunit 2; Interleukin-10 receptor subunit beta; RGD1560373 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.