Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL12B Antibody (OTI1A3), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB271032A48 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB271032A48 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB271032A48 Supplier Novus Biologicals Supplier No. NBP271032AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL12B Monoclonal specifically detects IL12B in Human samples. It is validated for Western Blot.

Specifications

Antigen IL12B
Applications Western Blot
Classification Monoclonal
Clone OTI1A3
Conjugate Alexa Fluor 488
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CLMF, CLMF p40, CLMF2, Cytotoxic lymphocyte maturation factor 40 kDa subunit, IL-12 subunit p40, IL-12BNK cell stimulatory factor chain 2, interleukin 12, p40, interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocytematuration factor 2, p40), interleukin-12 beta chain, interleukin-12 subunit beta, natural killer cell stimulatory factor, 40 kD subunit, NKSF, NKSF2IL12, subunit p40
Gene Symbols IL12B
Host Species Mouse
Immunogen Full length recombinant protein of human IL12B (NP_002178) produced in SF9 cell.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3593.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.