Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IL17RD Polyclonal Antibody
Rabbit Polyclonal Antibody
$589.50
Specifications
Antigen | IL17RD |
---|---|
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5111112
|
Invitrogen™
PA5111112 |
100 μL |
Each for $589.50
|
|
|||||
PIPA5111181
|
Invitrogen™
PA5111181 |
100 μL |
Each for $589.50
|
|
|||||
Description
Immunogen sequence: LACKPFWKPR NLNISQHGSD MQVSFDHAPH NFGFRFFYLH YKLKHEGPFK RKTCKQEQTT ETTSCLLQNV SPGDYIIELV DDTNTTRKVM.
Feedback inhibitor of fibroblast growth factor mediated Ras-MAPK signaling and ERK activation. May inhibit FGF-induced FGFR1 tyrosine phosphorylation. Regulates the nuclear ERK signaling pathway by spatially blocking nuclear translocation of activated ERK without inhibiting cytoplasmic phosphorylation of ERK. Mediates JNK activation and may be involved in apoptosis. Might have a role in the early stages of fate specification of GnRH-secreting neurons.Specifications
IL17RD | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NFM7 | |
54756 | |
Recombinant Protein corresonding to Human IL17RD. Recombinant protein control fragment (Product #RP-96358). | |
Antibody |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
Il17rd | |
2810004A10Rik; AI428510; HH18; hSef; IL-17 receptor D; IL17RD; IL-17RD; IL17Rhom; IL17RLM; interleukin 17 receptor D; interleukin-17 receptor D; Interleukin-17 receptor-like protein; mSef; RGD1565237; SEF; sef homolog; Sef-S; UNQ6115/PRO20026 | |
Il17rd | |
Primary | |
Antigen affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title