Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL18R1 Antibody (013), PerCP, Novus Biologicals™
SDP

Catalog No. NB333283 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB333283 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB333283 Supplier Novus Biologicals Supplier No. NBP290418PCP
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IL18R1 Monoclonal antibody specifically detects IL18R1 in Mouse samples. It is validated for ELISA

Specifications

Antigen IL18R1
Applications ELISA
Classification Monoclonal
Clone 013
Conjugate PerCP
Dilution ELISA
Formulation PBS
Gene Alias CD218 antigen-like family member A, CD218a, CD218a antigen, CDw218a, cytokine receptor, IL-18R1, IL-18R-1, IL1R-rp, IL1RRPIL1 receptor-related protein, IL-1RrpIL18RA, interleukin 18 receptor 1, interleukin-18 receptor 1
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Mouse IL18R1 (Accession#: NP_032391.1, Met 1-Gly 326)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline CD Markers
Primary or Secondary Primary
Gene ID (Entrez) 8809
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.