Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL18R1 Antibody (11), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB067980 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB067980 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB067980 Supplier Novus Biologicals Supplier No. NBP289797AF405
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IL18R1 Monoclonal specifically detects IL18R1 in Human samples. It is validated for ELISA.

Specifications

Antigen IL18R1
Applications ELISA
Classification Monoclonal
Clone 11
Conjugate Alexa Fluor 405
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias CD218 antigen-like family member A, CD218a, CD218a antigen, CDw218a, cytokine receptor, IL-18R1, IL-18R-1, IL1R-rp, IL1RRPIL1 receptor-related protein, IL-1RrpIL18RA, interleukin 18 receptor 1, interleukin-18 receptor 1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human IL18R1 (NP_003846.1; Met 1-Arg 329).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8809
Test Specificity No cross-reactivity in ELISA with: Mouse IL18R1/ IL1RRP/ CD218a Mouse SIGIRR Human IL1R1/ CD121a Human IL1R2/ IL1RB/ CD121b Human IL1R3/ IL1RAP Human IL1R8/ IL1RAPL1 Human IL1RL1/ IL1R4/ ST2 Human IL18RAP/ IL1R7 Human IL1R9/ IL1RAPL2
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.