Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL18R1 Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB089100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB089100 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB089100 Supplier Novus Biologicals Supplier No. NBP299706C

Rabbit Polyclonal Antibody

IL18R1 Polyclonal specifically detects IL18R1 in Human samples. It is validated for ELISA.

Specifications

Antigen IL18R1
Applications ELISA
Classification Polyclonal
Conjugate DyLight 650
Formulation 50 mM Sodium Borate
Gene Alias CD218 antigen-like family member A, CD218a, CD218a antigen, CDw218a, cytokine receptor, IL-18R1, IL-18R-1, IL1R-rp, IL1RRPIL1 receptor-related protein, IL-1RrpIL18RA, interleukin 18 receptor 1, interleukin-18 receptor 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant IL18R1 (NP_003846.1; Met 1-Arg 329).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8809
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.